missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(II) Chloride Dihydrate, 98%, ACS Reagent, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
12 kg
Packaging:
Drum with inliner
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Boiling Point | 652°C |
Melting Point | 37°C to 38°C |
Color | Colorless |
Quantity | 2.5 kg |
Linear Formula | SnCl2 · 2H2O |
UN Number | UN3260 |
Solubility Information | Soluble in water, Hydrochloric Acid |
Formula Weight | 225.65g/mol |
Percent Purity | 98% |
Grade | ACS Reagent |
Show More |
Safety and Handling



Product Identifier
- Tin(II) chloride dihydrate
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 C
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tin(II) Chloride Dihydrate, 98%, ACS Reagent, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction