missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(Ii) Chloride Dihydrate, Reagent grade, Honeywell™
Click to view available options
Quantity:
100 g
250 g
500 g
Description
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 500 g |
Linear Formula | SnCl2 · 2H2O |
UN Number | 3260 |
Solubility Information | Soluble in water |
Formula Weight | 225.65g/mol |
Percent Purity | 98% |
Grade | Reagent |
Chemical Name or Material | Tin(Ii) Chloride Dihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'