missing translation for 'onlineSavingsMsg'
Learn More
Tin(II) chloride dihydrate, Reagent Grade, Thermo Scientific Chemicals Product Code.: 11347227

Tin(II) chloride dihydrate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11347227
2 kg
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11347227

Brand: Thermo Scientific Alfa Aesar 011536.A3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Stannous chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-69-1
Molecular Formula SnCl2·2H2O
MDL Number MFCD00149863
Synonym Stannous chloride dihydrate

Specifications

Melting Point 43°C to 46°C
Physical Form Crystalline
Quantity 2 kg
Solubility Information Soluble in dilute and concentrated HCl,alcohol,ethyl acetate,glacial acetic acid,and NaOH solution. Decomposes in excess water.
Formula Weight 225.65 (189.62 Anhydrous)
Grade Reagent
Sensitivity Air Sensitive
Density 2.71 g/mL
Chemical Name or Material Tin(II) chloride dihydrate, Reagent Grade

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW06K063 Certificate TypeCertificate of Analysis Date13/09/2024 Catalog Number011536.A3,011536.36,011536.22
Lot NumberU24G005 Certificate TypeCertificate of Analysis Date26/04/2023 Catalog Number011536.A3,011536.36,011536.22
Lot NumberZ16G012 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number011536.A3,011536.36,011536.22
Lot NumberQ29I011 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog Number011536.A3,011536.36,011536.22
Lot NumberQ13I033 Certificate TypeCertificate of Analysis Date22/04/2022 Catalog Number011536.A3,011536.36,011536.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) chloride dihydrate, Reagent Grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.