missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(Ii) Chloride, Honeywell
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2 kg
Beschreibung
- ≥95% for general laboratory use.
Spezifikation
Spezifikation
Quantity | 500 g |
Linear Formula | SnCl2 |
UN Number | 3260 |
Formula Weight | 189.62g/mol |
Percent Purity | 98% |
Grade | Reagent |
Chemical Name or Material | Tin(Ii) Chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'