missing translation for 'onlineSavingsMsg'
Learn More
Tin(II) fluoride, 99%, Thermo Scientific Chemicals Product Code.: 10657494

Tin(II) fluoride, 99%, Thermo Scientific Chemicals

Product Code. 10657494
500 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10657494

Brand: Thermo Scientific Acros 308505000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-47-3
Molecular Formula F2Sn
Molecular Weight (g/mol) 156.69
MDL Number MFCD00042540

Specifications

Color White
Physical Form Powder, Crystals or Crystalline Powder
Quantity 500 g
Assay Percent Range 99%
Linear Formula SnF2
Merck Index 15, 8911
Solubility Information Solubility in water: 300- 390g/L. Other solubilities: insoluble in ethanol,diethyl ether,chloroform
Formula Weight 156.69
Percent Purity 99%
Packaging Plastic bottle
Chemical Name or Material Tin(II) fluoride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465802 Certificate TypeCertificate of Analysis Date09/09/2024 Catalog Number308500050,308505000,308501000
Lot NumberA0465394 Certificate TypeCertificate of Analysis Date03/09/2024 Catalog Number308500050,308505000,308501000
Lot NumberA0454445 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number308500050,308505000,308501000
Lot NumberA0456506 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number308500050,308505000,308501000
Lot NumberA0458907 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number308500050,308505000,308501000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) fluoride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.