missing translation for 'onlineSavingsMsg'
Learn More
Tin(II) fluorophosphate, Thermo Scientific Chemicals Product Code.: 11304547

Tin(II) fluorophosphate, Thermo Scientific Chemicals

Product Code. 11304547
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11304547

Brand: Thermo Scientific Alfa Aesar 039608.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52262-58-5
Molecular Formula FO3PSn
Molecular Weight (g/mol) 216.679
MDL Number MFCD00058794
InChI Key ZIQLCYCKKROENE-UHFFFAOYSA-L
Synonym tin ii fluorophosphate, stannous fluorophosphate, acmc-20ajhz, fluoridophosphoric acid tin ii salt, tin, ion sn2+ fluoro-phosphite ion, fluoro-dioxido-oxophosphorane; tin 2+, fluoranyl-bis oxidanidyl-oxidanylidene-$l^ 5-phosphane; tin 2+
PubChem CID 44717639
IUPAC Name fluoro-dioxido-oxo-$l^{5}-phosphane;tin(2+)
SMILES [O-]P(=O)([O-])F.[Sn+2]

Specifications

Color White
Physical Form Powder
Quantity 5 g
Solubility Information Slightly soluble (3.0g/L at 25°C)
Formula Weight 216.66
Chemical Name or Material Tin(II) fluorophosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) fluorophosphate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.