missing translation for 'onlineSavingsMsg'
Learn More
Tin(II) iodide, 99+%, Thermo Scientific Chemicals Product Code.: 11304809

Tin(II) iodide, 99+%, Thermo Scientific Chemicals

Product Code. 11304809
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11304809

Brand: Thermo Scientific Alfa Aesar 071112.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tin(II) iodide is used to prepare organic-inorganic perovskite thin films which exhibit uniform surfaces and strong photoluminescence. It finds application as an analytical reagent in the determination of rhodium and iridium by spectrophotometry. It is also involved in the synthesis of CH3(CH2)11NH3]SnI3, which is a hybrid semiconductor with MoO3 type tin(II) iodide layers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-70-9
Molecular Formula I2Sn
Molecular Weight (g/mol) 372.519
MDL Number MFCD00049545
InChI Key JTDNNCYXCFHBGG-UHFFFAOYSA-L
Synonym tin ii iodide, stannous iodide, tin diiodide, tin iodide sni2, stannous diiodide, sni2, acmc-1bqot, tin ii iodide, ultra dry, tin ii iodide,-10 mesh trace metals basis, tin ii iodide, anhydrous, beads,-10 mesh trace metals basisShow MoreShow Less
PubChem CID 25138
IUPAC Name diiodotin
SMILES [Sn](I)I

Specifications

Boiling Point 720°C (decomposition)
Melting Point 320°C
Physical Form Powder
Quantity 50 g
Assay Percent Range >99%
Solubility Information Soluble in benzene,carbon disulfide,chloroform,and solutions of alkali chlorides and iodides. Decomposes in water
Formula Weight 372.5
Sensitivity Air, light, and moisture sensitive
Density 5.28
Chemical Name or Material Tin(II) iodide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP07L064 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog Number071112.09,071112.18
Lot NumberY22J016 Certificate TypeCertificate of Analysis Date01/01/2024 Catalog Number071112.09,071112.18
Lot NumberQ20I018 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number071112.09,071112.18
Lot NumberZ11F033 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number071112.09,071112.18
Lot NumberH25U019 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number071112.09,071112.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) iodide, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.