missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) oxalate, Thermo Scientific Chemicals

Product Code. 11307717
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tin(II) oxalate is used for dyeing and printing textiles. It acts as a catalyst for the esterification reactions between long chain fatty acids and alcohols and in polymerization reactions. It is also used in stannous oral care compositions. Further, it is used in proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-94-8
Molecular Formula C2O4Sn
Molecular Weight (g/mol) 206.728
MDL Number MFCD00040678
InChI Key OQBLGYCUQGDOOR-UHFFFAOYSA-L
Synonym stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, sar72fe8eh, stavelan cinaty
PubChem CID 13149
IUPAC Name oxalate;tin(2+)
SMILES C(=O)(C(=O)[O-])[O-].[Sn+2]

Specifications

Melting Point 280°C (decomposition)
Color White
Density 3.56 g/mL
Linear Formula SnC2O4
Quantity 500 g
UN Number UN3288
Beilstein 3708588
Merck Index 14,8786
Solubility Information Soluble in dilute HCl; Insoluble in water;Soluble in acids; Insoluble in water and acetone.
Formula Weight 206.72
Physical Form Powder
Chemical Name or Material Tin(II) oxalate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'