missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) oxalate, Thermo Scientific Chemicals

£25.23 - £94.97

Chemical Identifiers

CAS 814-94-8
Molecular Formula C2O4Sn
Molecular Weight (g/mol) 206.728
MDL Number MFCD00040678
InChI Key OQBLGYCUQGDOOR-UHFFFAOYSA-L
Synonym stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, sar72fe8eh, stavelan cinaty
PubChem CID 13149
IUPAC Name oxalate;tin(2+)
SMILES C(=O)(C(=O)[O-])[O-].[Sn+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11397707
View Documents
Thermo Scientific Alfa Aesar
014113.22
100 g
£25.23
100g
Please to purchase this item. Need a web account? Register with us today!
11307717
View Documents
Thermo Scientific Alfa Aesar
014113.36
500 g
£94.97
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tin(II) oxalate is used for dyeing and printing textiles. It acts as a catalyst for the esterification reactions between long chain fatty acids and alcohols and in polymerization reactions. It is also used in stannous oral care compositions. Further, it is used in proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

814-94-8
206.728
OQBLGYCUQGDOOR-UHFFFAOYSA-L
13149
C(=O)(C(=O)[O-])[O-].[Sn+2]
C2O4Sn
MFCD00040678
stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, sar72fe8eh, stavelan cinaty
oxalate;tin(2+)

Specifications

814-94-8
White
C2O4Sn
MFCD00040678
3708588
stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, sar72fe8eh, stavelan cinaty
OQBLGYCUQGDOOR-UHFFFAOYSA-L
oxalate;tin(2+)
13149
Powder
280°C (decomposition)
3.56 g/mL
SnC2O4
UN3288
14,8786
Soluble in dilute HCl; Insoluble in water;Soluble in acids; Insoluble in water and acetone.
C(=O)(C(=O)[O-])[O-].[Sn+2]
206.728
206.72
Tin(II) oxalate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 212-414-0

missing translation for 'rtecsNumber' : XQ3950000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only