missing translation for 'onlineSavingsMsg'
Learn More
Tin(II) oxide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11317307

Tin(II) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11317307
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11317307

Brand: Thermo Scientific Alfa Aesar 011569.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reducing agent, soft abrasive, preparation of stannous salts

It is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21651-19-4
Molecular Formula OSn
Molecular Weight (g/mol) 134.71
MDL Number MFCD00011243
InChI Key QHGNHLZPVBIIPX-UHFFFAOYSA-N
Synonym stannous oxide, tin ii oxide, tin oxide sno, tin monoxide, tin oxide sn2o2, stannane, oxo, stannanone, stannicoxide, oxostannanylidene, oxo stannaneShow MoreShow Less
PubChem CID 88989
IUPAC Name oxotin
SMILES O=[Sn]

Specifications

Melting Point 1080°C (decomposition)
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Insoluble in water,and alcohol.
Formula Weight 134.69
Density 6.45 g/mL
Chemical Name or Material Tin(II) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX17K021 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog Number011569.22,011569.14,011569.36
Lot NumberX17F021 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011569.22,011569.14,011569.36
Lot NumberG05Z010 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011569.22,011569.14,011569.36
Lot NumberP11B003 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011569.22,011569.14,011569.36
Lot NumberL22K19 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011569.22,011569.14,011569.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) oxide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.