Learn More
Tin(II) oxide, 99%, Thermo Scientific Chemicals
Description
Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.
Solubility
Insoluble in water and alcohol.
Notes
Incompatible with materials like strong oxidizing agents, strong acids, strong reducing agents and alkali metals.
Specifications
Specifications
Melting Point | 1080°C (decomposition) |
Quantity | 500 g |
Linear Formula | SnO |
Merck Index | 14,8787 |
Solubility Information | Insoluble in water and alcohol. |
Formula Weight | 134.69 |
Percent Purity | 99% |
Odor | Odorless |
Density | 6.45 |
Chemical Name or Material | Tin(II) oxide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10224008 | Certificate of Analysis | 03/10/2024 | A17889.36,A17889.22,A17889.0E |
10249417 | Certificate of Analysis | 19/01/2024 | A17889.36,A17889.22,A17889.0E |
10242871 | Certificate of Analysis | 17/08/2022 | A17889.36,A17889.22,A17889.0E |
10155031 | Certificate of Analysis | 23/11/2021 | A17889.36,A17889.22,A17889.0E |
10224007 | Certificate of Analysis | 23/11/2021 | A17889.36,A17889.22,A17889.0E |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.