missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) pyrophosphate, Thermo Scientific Chemicals

£30.63 - £91.46

Chemical Identifiers

CAS 15578-26-4
Molecular Formula O7P2Sn2
Molecular Weight (g/mol) 411.361
MDL Number MFCD00049544
InChI Key GEZAUFNYMZVOFV-UHFFFAOYSA-J
Synonym stannous pyrophosphate, technescan pyp, ditin pyrophosphate, tin ii pyrophosphate, tin 2+ pyrophosphate, unii-4dnt29ec86, stannous pyrophosphate usan, tin 2+ diphosphate 1:2, pyro stannous phosphate, ditin 2+ pyrophosphate 4-Show MoreShow Less
PubChem CID 66379
IUPAC Name phosphonato phosphate;tin(2+)
SMILES [O-]P(=O)([O-])OP(=O)([O-])[O-].[Sn+2].[Sn+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11403830
SDS View Documents
Thermo Scientific Alfa Aesar
089785.18
50 g
£30.63
50g
Please to purchase this item. Need a web account? Register with us today!
11413830
SDS View Documents
Thermo Scientific Alfa Aesar
089785.30
250 g
£91.46
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tin(II) pyrophosphate is used to inhibit the formation of volatile sulfur compounds. Further, it is used for cyanide-free tin coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.

Irritating to eyes, respiratory system and skin.

missing translation for 'einecsNumber' : 239-635-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) pyrophosphate, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.