missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) sulfate, 95.5% min, Thermo Scientific Chemicals

Product Code. 11387227
250 g
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in tin plating baths and it is a precursor for tin metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7488-55-3
Molecular Formula O4SSn
Molecular Weight (g/mol) 214.77
MDL Number MFCD00011246
InChI Key OBBXFSIWZVFYJR-UHFFFAOYSA-L
Synonym stannous sulfate, tin ii sulfate, tin 2+ sulfate, unii-0mfe10j96e, sulfuric acid, tin 2+ salt 1:1, stannous sulfate, crystal, stannous sulfate, solutions, tin, ion sn2+ sulfate, tin ii sulfate 250g
PubChem CID 62643
IUPAC Name tin(2+);sulfate
SMILES [Sn++].[O-]S([O-])(=O)=O

Specifications

Melting Point 378°C (decomposition)
Physical Form Powder
Quantity 250 g
Assay Percent Range 95.5% min.
Solubility Information Soluble in water and dilute H2SO4.
Formula Weight 214.77
Density 4.15 g/mL
Chemical Name or Material Tin(II) sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) sulfate, 95.5% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.