missing translation for 'onlineSavingsMsg'
Learn More
Tin(II) sulfide, 99.5%, Thermo Scientific Chemicals Product Code.: 11317667

Tin(II) sulfide, 99.5%, Thermo Scientific Chemicals

Product Code. 11317667
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11317667

Brand: Thermo Scientific Alfa Aesar 014051.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polymerization catalyst.Tin(II) sulfide is used as a polymerization catalyst. It is also used as a friction material. Further, it is used in optoelectronic device as well as a component in heterojunction photovoltaic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-95-0
Molecular Formula SSn
Molecular Weight (g/mol) 150.77
MDL Number MFCD00011245
InChI Key DZXKSFDSPBRJPS-UHFFFAOYSA-N
Synonym tin ii sulfide, stannous sulfide, tin sulfide sns, tin monosulfide, tin sulphide, stannanethione, herzenbergite, zinnsulfur, acmc-1btyn
PubChem CID 426379
IUPAC Name λ²-tin(2+) sulfanediide
SMILES [S--].[Sn++]

Specifications

Melting Point 882°C
Quantity 5 g
Assay Percent Range 99.5%
Solubility Information Soluble in conc. HCl,hot conc. H2SO4.Insoluble in water.
Formula Weight 150.75
Density 5.08
Chemical Name or Material Tin(II) sulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) sulfide, 99.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.