Learn More
Tin(II) tartrate hydrate, 95%, Thermo Scientific Chemicals
£100.50 - £375.87
Chemical Identifiers
CAS | 815-85-0 |
---|---|
Molecular Formula | C4H4O6Sn |
Molecular Weight (g/mol) | 266.78 |
MDL Number | MFCD00054358 |
InChI Key | YXTDAZMTQFUZHK-UHFFFAOYNA-L |
Synonym | unii-6js9u5611x, stannous tartrate, tin tartrate, tartaric acid, tin 2+ salt 1:1, r-r*,r*-2,3-dihydroxybutanedioic acid, tin r-r*,r*-tartrate, butanedioic acid, 2,3-dihydroxy-, r-r*,r*-, tin 2+ salt 1:1, butanedioic acid, 2,3-dihydroxy-theta-theta,theta-, tin 2++ salt 1:1Show More |
PubChem CID | 66545835 |
IUPAC Name | (2R,3R)-2,3-dihydroxybutanedioate;tin(2+) |
SMILES | [Sn++].OC(C(O)C([O-])=O)C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11304537
|
Thermo Scientific Alfa Aesar
039604.14 |
25 g |
£100.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314537
|
Thermo Scientific Alfa Aesar
039604.22 |
100 g |
£375.87
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used for dyeing and printing textiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
815-85-0 | |
266.78 | |
YXTDAZMTQFUZHK-UHFFFAOYNA-L | |
66545835 | |
[Sn++].OC(C(O)C([O-])=O)C([O-])=O |
C4H4O6Sn | |
MFCD00054358 | |
unii-6js9u5611x, stannous tartrate, tin tartrate, tartaric acid, tin 2+ salt 1:1, r-r*,r*-2,3-dihydroxybutanedioic acid, tin r-r*,r*-tartrate, butanedioic acid, 2,3-dihydroxy-, r-r*,r*-, tin 2+ salt 1:1, butanedioic acid, 2,3-dihydroxy-theta-theta,theta-, tin 2++ salt 1:1Show More | |
(2R,3R)-2,3-dihydroxybutanedioate;tin(2+) |
Specifications
815-85-0 | |
C4H4O6Sn | |
14,8792 | |
slightly soluble in water. | |
[Sn++].OC(C(O)C([O-])=O)C([O-])=O | |
266.78 | |
266.76 (Anhydrous) | |
Odorless | |
Tin(II) tartrate hydrate |
Powder | |
MFCD00054358 | |
unii-6js9u5611x, stannous tartrate, tin tartrate, tartaric acid, tin 2+ salt 1:1, r-r*,r*-2,3-dihydroxybutanedioic acid, tin r-r*,r*-tartrate, butanedioic acid, 2,3-dihydroxy-, r-r*,r*-, tin 2+ salt 1:1, butanedioic acid, 2,3-dihydroxy-theta-theta,theta-, tin 2++ salt 1:1 | |
YXTDAZMTQFUZHK-UHFFFAOYNA-L | |
(2R,3R)-2,3-dihydroxybutanedioate;tin(2+) | |
66545835 | |
95% | |
Air Sensitive |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 212-427-1
missing translation for 'rtecsNumber' : EJ9892500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Tin(II) tartrate hydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Tin(II) tartrate hydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.