missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) telluride, 99.999% (metals basis)

Product Code. 11319409
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11319409

Brand: Thermo Scientific Alfa Aesar 089186.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12040-02-7 | H2SnTe | 248.33 g/mol

Tin(II) telluride is used as detector material for photodiodes in infrared used spectral range.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12040-02-7
Molecular Formula H2SnTe
Molecular Weight (g/mol) 248.33
MDL Number MFCD00049543
InChI Key UURRKPRQEQXTBB-UHFFFAOYSA-N
IUPAC Name stannanetellone
SMILES [SnH2]=[Te]

Specifications

Melting Point 780°C
Physical Form Pieces
Quantity 10 g
UN Number UN3288
Solubility Information Insoluble in water.
Formula Weight 246.29
Percent Purity 99.999%
Odor Odorless
Density 6.48 g/mL
Assay (metals basis)
Chemical Name or Material Tin (II) telluride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.