missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) 2-ethylhexanoate diisopropoxide, 98% (metals basis), Thermo Scientific Chemicals

£33.30 - £434.00

Chemical Identifiers

CAS 288374-32-3
MDL Number MFCD00210604
Synonym tin iv 2-ethylhexanoate diisopropoxide, tin iv 2-ethylhexanoatediisopropoxide metalsbasis
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11303548
View Documents
Thermo Scientific Alfa Aesar
039169.03
1 g
£33.30
1g
Please to purchase this item. Need a web account? Register with us today!
11393538
View Documents
Thermo Scientific Alfa Aesar
039169.06
5 g
£136.00
5g
Please to purchase this item. Need a web account? Register with us today!
11313548
View Documents
Thermo Scientific Alfa Aesar
039169.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

288374-32-3
MFCD00210604
tin iv 2-ethylhexanoate diisopropoxide, tin iv 2-ethylhexanoatediisopropoxide metalsbasis

Specifications

288374-32-3
Moisture sensitive
523.08
Liquid
Tin(IV) 2-ethylhexanoate diisopropoxide
MFCD00210604
tin iv 2-ethylhexanoate diisopropoxide, tin iv 2-ethylhexanoatediisopropoxide metalsbasis
98%
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H315-H319
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.

P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H361d

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only