missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) acetate, 98%, extra pure, Thermo Scientific Chemicals

Product Code. 10431913
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2800-96-6
Molecular Formula C8H12O8Sn
Molecular Weight (g/mol) 354.89
MDL Number MFCD00014976
InChI Key YJGJRYWNNHUESM-UHFFFAOYSA-J
Synonym tin iv acetate, tin acetate, tetraacetoxystannane, tin-iv-acetate, acmc-20ajhv, tin 4+ ion tetraacetate, tetraacetoxytin, tin 4+ tetraacetate
PubChem CID 9863446
IUPAC Name tin(4+);tetraacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sn+4]

Specifications

Melting Point 231.0°C to 237.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 98%
Linear Formula (CH3COO)4Sn
Beilstein 02, III, 134
Solubility Information Solubility in water: hydrolysis. Other solubilities: hydroylsis with water to sn(iv) oxide hydrate and, acetic acid;, soluble in acetone, benzene;, less soluble in carbon tetrachloride
Formula Weight 354.89
Percent Purity 98%
Grade Extra Pure
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Tin(IV) acetate
Show More Show Less
compliance-icons
Nombre del producto
  • Tin(IV) acetate
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
Indicaciones de peligro
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH044-Risk of explosion if heated under confinement.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'