missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 378201000

37.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.



 View more versions of this product

Product Code. 10629784

  • £47.24 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tin(IV) chloride
7646-78-8, 75-09-2
Cl4Sn
09,436; 17,335; 14,304; 13,300; 12,486; 10,370; 07,342; 06,553; 05,627; 03,269; 01,1111; 11,552
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
HPGGPRDJHPYFRM-UHFFFAOYSA-J
tetrachlorostannane
24287
1M, exact strength on the certificate of analysis
Clear colorless solution, fuming in air
1.4190g/mL
1M Solution in Dichloromethane
100 mL
MFCD00011242
15, 8901
Solubility in water: reacts
Cl[Sn](Cl)(Cl)Cl
260.52
260.52
AcroSeal™ Glass bottle
1.419
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tin(IV) chloride, 1M solution in dichloromethane

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only