Learn More
Tin(IV) chloride, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Tin(IV) chloride is a precursor to prepare organotin compounds such as tetralkyltin and dialkyldichlorotin(IV), which finds application as catalysts and polymer stabilizers. As a Lewis acid catalyst, it is used in Fridel-Crafts reactions for alkylation and cyclization. It is involved in the selective nitration of aromatic compounds in presence of fuming nitric acid. Furthermore, it is used to prepare tin(IV) oxide coating by sol-gel process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 114°C |
Melting Point | °C to 33°C |
Physical Form | Liquid |
Quantity | 25 g |
Assay Percent Range | 99.99% (metals basis) |
Solubility Information | Miscible with alcohol,benzene,toluene,chloroform,acetone,carbon terachloride,gasoline and carbon disulfide. |
Formula Weight | 260.5 |
Refractive Index | 1.512 |
Sensitivity | Moisture sensitive |
Density | 2.26 g/mL |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T25J039 | Certificate of Analysis | 07/08/2023 | 071203.14,071203.22 |
Z06I018 | Certificate of Analysis | 09/01/2023 | 071203.14,071203.22 |
P24G009 | Certificate of Analysis | 26/09/2022 | 071203.14,071203.22 |
Y23H015 | Certificate of Analysis | 12/04/2022 | 071203.14,071203.22 |
K19T013 | Certificate of Analysis | 08/12/2021 | 071203.14,071203.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.