missing translation for 'onlineSavingsMsg'
Learn More
Tin(IV) chloride, 99%, anhydrous, Thermo Scientific Chemicals Product Code.: 10472391

Tin(IV) chloride, 99%, anhydrous, Thermo Scientific Chemicals

Product Code. 10472391
10 mL, Glass bottle
Click to view available options
Quantity:
10 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10472391

Brand: Thermo Scientific Acros 197510100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-78-8
Molecular Formula Cl4Sn
Molecular Weight (g/mol) 260.52
InChI Key HPGGPRDJHPYFRM-UHFFFAOYSA-J
Synonym tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
PubChem CID 24287
IUPAC Name tetrachlorostannane
SMILES Cl[Sn](Cl)(Cl)Cl

Specifications

Boiling Point 114.0°C
Melting Point -33.0°C
Color Brown-Red
Physical Form Solution
Quantity 10 mL
Assay Percent Range 98.5 to 101.5% (Argentometry)
Linear Formula SnCl4
Fieser 01,1111; 03,269; 05,627; 06,553; 07,342; 09,436; 11,522; 12,486; 13,14; 14,304; 15,311; 17,335
Merck Index 15,8901
Solubility Information Solubility in water: reacts. Other solubilities: soluble in carbon disulfide, alcohol, carbon, tetrachloride, benzene, toluene, acetone, kerosene, and gasoline
Formula Weight 260.52
Percent Purity 99%
Packaging Glass bottle
Specific Gravity 2.22
Density 2.2200g/mL
Chemical Name or Material Tin(IV) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466182 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog Number197510100,197510025,197510010,197511000
Lot NumberA0464755 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number197510100,197510025,197510010,197511000
Lot NumberA0462976 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog Number197510100,197510025,197510010,197511000
Lot NumberA0457802 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number197510100,197510025,197510010,197511000
Lot NumberA0456203 Certificate TypeCertificate of Analysis Date08/09/2023 Catalog Number197510100,197510025,197510010,197511000
compliance-icons compliance-icons
Product Identifier
  • Tin(IV) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(IV) chloride, 99%, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.