missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride, anhydrous, 98% (metals basis), Thermo Scientific Chemicals

£80.90 - £286.43

Chemical Identifiers

CAS 7646-78-8
Molecular Formula Cl4Sn
Molecular Weight (g/mol) 260.51
MDL Number MFCD00011242
InChI Key HPGGPRDJHPYFRM-UHFFFAOYSA-J
Synonym tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
PubChem CID 24287
IUPAC Name tetrachlorostannane
SMILES Cl[Sn](Cl)(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15495975
View Documents
Thermo Scientific Alfa Aesar
011571.36
500 g
£80.90
500g
Please to purchase this item. Need a web account? Register with us today!
15405985
View Documents
Thermo Scientific Alfa Aesar
011571.57
4 x 500 g
£286.43
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tin(IV) chloride is a precursor to prepare organotin compounds such as tetralkyltin and dialkyldichlorotin(IV), which find applications as catalysts and polymer stabilizers. As a Lewis acid catalyst, it is used in Fridel-Crafts reactions for alkylation and cyclization. It is involved in the selective nitration of aromatic compounds in the presence of fuming nitric acid. Furthermore, it is used to prepare tin(IV) oxide coating by sol-gel process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7646-78-8
260.51
HPGGPRDJHPYFRM-UHFFFAOYSA-J
24287
Cl[Sn](Cl)(Cl)Cl
Cl4Sn
MFCD00011242
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
tetrachlorostannane

Specifications

114°C
7646-78-8
Liquid
SnCl4
UN1827
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
HPGGPRDJHPYFRM-UHFFFAOYSA-J
tetrachlorostannane
24287
98%
Moisture sensitive
(metals basis)
-33°C
Colorless
Cl4Sn
MFCD00011242
14,8774
Soluble in alcohol,CCl4,benzene,toluene,chloroform,carbon terachloride,gasoline,carbon disulfide and acetone; Soluble in water with much heat evolution.
Cl[Sn](Cl)(Cl)Cl
260.51
260.5
1.512
2.26 g/mL
Tin(IV) chloride, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H330-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: STANNIC CHLORIDE, ANHYDROUS

missing translation for 'einecsNumber' : 231-588-9

missing translation for 'rtecsNumber' : XP8750000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only