missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride bis(2,4-pentanedionate), 95%, Thermo Scientific Chemicals

Product Code. 11367298
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a raw material in synthesis. It is also used as NMR shift reagents and as catalysts for organic synthesis, and precursors to industrial hydroformylation catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16919-46-3
Molecular Formula C10H14Cl2O4Sn
Molecular Weight (g/mol) 387.83
MDL Number MFCD00015316
InChI Key DTMARTPDQMQTRX-VGKOASNMSA-J
Synonym Bis(acetylacetonato)dichlorotin(IV); Tin(IV) bis(acetylacetonate) chloride
SMILES [Cl-].[Cl-].[Sn+4].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 199°C
Physical Form Crystalline
Quantity 10 g
UN Number UN3146
Solubility Information Soluble in organic solvents.
IUPAC Name tin(4+) bis((2Z)-4-oxopent-2-en-2-olate) dichloride
Formula Weight 387.83
Percent Purity 95%
Sensitivity Moisture sensitive
Chemical Name or Material Tin(IV) chloride bis(2,4-pentanedionate)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'