missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) chloride hydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Tin(IV) chloride hydrate is used in the formation of a xerogel via a cyanogel, a gel that has potential use in solid-state gas sensors. It is involved in the preparation of Indium tin oxide thin films by chemical vapor deposition method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 56°C |
Physical Form | Lump |
Quantity | 100 g |
Assay Percent Range | 98% |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 260.50 (Anhydrous) |
Chemical Name or Material | Tin(IV) chloride hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'