missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride pentahydrate, 98+%, extra pure, Thermo Scientific Chemicals

£30.65 - £88.34

Chemical Identifiers

CAS 10026-06-9
Molecular Formula Cl4H10O5Sn
Molecular Weight (g/mol) 350.59
MDL Number MFCD00149864
InChI Key KHMOASUYFVRATF-UHFFFAOYSA-J
Synonym tin iv chloride pentahydrate, tetrachlorostannane pentahydrate, tin chloride pentahydrate, stannic chloride pentahydrate, stannane, tetrachloro-, pentahydrate, ccris 6330, tin iv chloride, pentahydrate 1:4:5, cl4sn.5h2o, stannic chloride 5h2o
PubChem CID 61442
IUPAC Name tin(4+) pentahydrate tetrachloride
SMILES O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10751691
View Documents Promotion Details
Thermo Scientific Acros
223691000
100 g Glass bottle
£30.65
100g
Please to purchase this item. Need a web account? Register with us today!
10147793
View Documents Promotion Details
Thermo Scientific Acros
223695000
500 g Glass bottle
£88.34
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Chemical Identifiers

10026-06-9
350.59
KHMOASUYFVRATF-UHFFFAOYSA-J
61442
O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]
Cl4H10O5Sn
MFCD00149864
tin iv chloride pentahydrate, tetrachlorostannane pentahydrate, tin chloride pentahydrate, stannic chloride pentahydrate, stannane, tetrachloro-, pentahydrate, ccris 6330, tin iv chloride, pentahydrate 1:4:5, cl4sn.5h2o, stannic chloride 5h2o
tin(4+) pentahydrate tetrachloride

Specifications

56.0°C
Colorless to Yellow
98% min
SnCl4·5H2O
14, 8774
Solubility in water: soluble. Other solubilities: soluble in alcohol
O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]
350.59
350.57
Extra Pure
10026-06-9
Liquid
Cl4H10O5Sn
MFCD00149864
tin iv chloride pentahydrate, tetrachlorostannane pentahydrate, tin chloride pentahydrate, stannic chloride pentahydrate, stannane, tetrachloro-, pentahydrate, ccris 6330, tin iv chloride, pentahydrate 1:4:5, cl4sn.5h2o, stannic chloride 5h2o
KHMOASUYFVRATF-UHFFFAOYSA-J
tin(4+) pentahydrate tetrachloride
61442
≥98%
Tin(IV) chloride pentahydrate, Extra Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only