missing translation for 'onlineSavingsMsg'
Learn More

Tin(Iv) Chloride Pentahydrate, Pure, Fisher Chemical™

Product Code. 10306300 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10306300

Brand: Fisher Chemical T/1661/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10026-06-9
Molecular Formula Cl4H10O5Sn
Molecular Weight (g/mol) 350.59
MDL Number MFCD00149864
InChI Key KHMOASUYFVRATF-UHFFFAOYSA-J
Synonym tin iv chloride pentahydrate, tetrachlorostannane pentahydrate, tin chloride pentahydrate, stannic chloride pentahydrate, stannane, tetrachloro-, pentahydrate, ccris 6330, tin iv chloride, pentahydrate 1:4:5, cl4sn.5h2o, stannic chloride 5h2o
PubChem CID 61442
IUPAC Name tin(4+) pentahydrate tetrachloride
SMILES O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]

Specifications

Melting Point 56°C
Color White
pH 2
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 350.57g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Tin(IV) chloride pentahydrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.