missing translation for 'onlineSavingsMsg'
Learn More
Tin(IV) iodide, 99+%, -10 mesh, Thermo Scientific Chemicals Product Code.: 10434325

Tin(IV) iodide, 99+%, -10 mesh, Thermo Scientific Chemicals

Product Code. 10434325
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10434325

Brand: Thermo Scientific Acros 388560500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-47-8
Molecular Formula H4I4Sn
Molecular Weight (g/mol) 630.36
MDL Number MFCD00036272
InChI Key ZBCVLXKKLMNLGV-UHFFFAOYSA-N
Synonym stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide
PubChem CID 24631
IUPAC Name tetraiodostannane
SMILES [Sn].I.I.I.I

Specifications

Boiling Point 364°C
Melting Point 144°C
Color Brown
Physical Form Powder
Quantity 50 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula SnI4
Merck Index 15, 8904
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,benzene,chloroform,ether and,carbon disulfide
Formula Weight 626.31
Percent Purity ≥99%
Packaging Glass bottle
Chemical Name or Material Tin(IV) iodide
Show More Show Less
compliance-icons
Product Identifier
  • Tin(IV) iodide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(IV) iodide, 99+%, -10 mesh, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.