Learn More
Tin(IV) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
£231.15 - £671.34
Chemical Identifiers
CAS | 7790-47-8 |
---|---|
Molecular Formula | H4I4Sn |
Molecular Weight (g/mol) | 630.36 |
MDL Number | MFCD00036272 |
InChI Key | ZBCVLXKKLMNLGV-UHFFFAOYSA-N |
Synonym | stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide |
PubChem CID | 24631 |
IUPAC Name | tetraiodostannane |
SMILES | [Sn].I.I.I.I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366878
|
Thermo Scientific Alfa Aesar
013617.06 |
5 g |
£231.15
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356878
|
Thermo Scientific Alfa Aesar
013617.14 |
25 g |
£671.34
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tin(IV) iodide is used in the preparation of tin(IV) oxide and tin(II) oxide. It reacts with hydroiodic acid to get rare tin hexaiodide. It forms tetraiodobis triphenylphosphine tin complexes by reacting with triphenylphosphine, which finds application in toothpaste formulation. Further, it acts as analytical reagents and also used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
7790-47-8 | |
630.36 | |
ZBCVLXKKLMNLGV-UHFFFAOYSA-N | |
24631 | |
[Sn].I.I.I.I |
H4I4Sn | |
MFCD00036272 | |
stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide | |
tetraiodostannane |
Specifications
340°C | |
7790-47-8 | |
H4I4Sn | |
UN3260 | |
stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide | |
ZBCVLXKKLMNLGV-UHFFFAOYSA-N | |
tetraiodostannane | |
24631 | |
99.998% | |
2.106 | |
4.46 g/mL | |
Tin(IV) iodide, Ultra dry |
∼143°C | |
Powder | |
MFCD00036272 | |
14,8777 | |
Soluble in alcohol,benzene,carbon disulfide,chloroform and ether. | |
[Sn].I.I.I.I | |
630.36 | |
626.31 | |
Ampule | |
Moisture sensitive | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 232-208-4
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only