missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Description
Tin(IV) isopropoxide isopropanol is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, thin films doped tin oxide nanoparticles, thin films and are widely studied as gas sensors. Tin(IV) precursor solution was prepared from tin(IV) isopropoxide dissolved in isopropyl alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Quantity | 5 mL |
UN Number | UN1993 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in hydrocarbons,reacts with alcohols,ketones and esters |
Formula Weight | 355.06 |
Percent Purity | 98% |
Physical Form | Liquid |
Assay | (metals basis) |
Chemical Name or Material | Tin(IV) isopropoxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'