missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals

Product Code. 15476945
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Tin(IV) isopropoxide is used as a precursor for the preparation of tin oxide nanoparticles, thin films, indium tin oxide (ITO) films , which shows desired electrochemical and conducting properties. Doped tin oxide nanoparticles and thin films find application as gas sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-61-8
Molecular Formula C12H32O4Sn
Molecular Weight (g/mol) 359.094
MDL Number MFCD00145408
InChI Key VUDAJANAMZGVOP-UHFFFAOYSA-N
Synonym tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate
PubChem CID 85354423
IUPAC Name propan-2-ol;tin
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn]

Specifications

Color Colorless
Odor Alcohol-like
Linear Formula Sn[OCH(CH3)2]4
Quantity 5 mL
UN Number UN3274
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohols,ketones and esters.
Formula Weight 355.06
Percent Purity 99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tin (IV) isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'