missing translation for 'onlineSavingsMsg'
Learn More
Tin(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11383378

Tin(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11383378
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11383378

Brand: Thermo Scientific Alfa Aesar 036302.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For vacuum deposition. Used as a catalyst, in putty, as a polishing powder for steel and glass, in ceramic glazes and colors. The electrical properties of tin (iv) oxide render it an excellent material for the detection of very low levels of carbon monoxide, and several devices based on sintered pellets of SnO 2 are commercially available. Dye-sensitized fully solid state cells having the structure of rough n-type semiconductor film comprising zinc and tin (IV) oxide/Ru-bipyridyl complex/p-CuI are found to generate high short-circuit photocurrents and open-circuit voltages. The preparation of hydrous tin (IV) oxide in a suitable form for use as an ion exchanger is described. a colloidal-crystal templating technique to prepare three-dimensionally ordered macroporous (3DOM) tin (iv) oxide films for use as lithium ion anodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydrideShow MoreShow Less
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name dioxotin
SMILES O=[Sn]=O

Specifications

Boiling Point 1800°C to 1900°C (sublimation)
Melting Point 1630°C
Color White
Physical Form Sintered Lump
Quantity 100 g
Linear Formula SnO2
Merck Index 14,8778
Solubility Information Insoluble in water. Soluble in concentrated sulfuric acid,hydrochloric acid. Slowly soluble in hot concentrated potassium or sodium hydroxide solution.
Formula Weight 150.69
Percent Purity 99.9%
Odor Odorless
Refractive Index 1.9968
Density 6.95 g/mL
Assay (metals basis)
Chemical Name or Material Tin(IV) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.