missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
It is employed in the manufacture of white enamels and for polishing glass. It has a broad range of applications in many areas such as semiconductors, photovoltaic materials, solar cells, lithium ion batteries, photocatalysts, and as gas-sensing agents in solid-state detectors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 1800°C to 1900°C (sublimation) |
Melting Point | 1630°C |
Color | White to Gray |
Physical Form | Powder |
Quantity | 2 kg |
Assay Percent Range | 99.9% metal basis |
Solubility Information | Soluble in hot concentrated alkalis,and concentrated acids. Insoluble in alcohol. |
IUPAC Name | stannanedione |
Formula Weight | 150.69 |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'