missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) oxide, 99.9%, (trace metal basis)

Product Code. 10637382 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10637382

Brand: Thermo Scientific Acros 223685000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18282-10-5 | O2Sn | 150.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name stannanedione
SMILES O=[Sn]=O

Specifications

Color White to Gray
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.9%
Linear Formula SnO2
Merck Index 15, 8905
Solubility Information Solubility in water: insoluble. Other solubilities: slowly soluble in sodium hydroxide solution, insoluble in alcohol and cold acids
Formula Weight 150.69
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Plastic bottle
Specific Gravity 6.95
Density 6.9500g/mL
Chemical Name or Material Tin(IV) oxide, 99.9%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.