missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) oxide, 99%, pure, Thermo Scientific Chemicals

£19.33 - £197.99

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name dioxotin
SMILES O=[Sn]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15575126
SDS View Documents
Thermo Scientific Acros
451811000
100 g
£19.33
100g
Please to purchase this item. Need a web account? Register with us today!
15555126
SDS View Documents
Thermo Scientific Acros
451810010
1 kg
£87.54
1kg
Please to purchase this item. Need a web account? Register with us today!
15565126
SDS View Documents
Thermo Scientific Acros
451810025
2.5 kg
£197.99
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18282-10-5
150.71
XOLBLPGZBRYERU-UHFFFAOYSA-N
29011
dioxotin
O2Sn
MFCD00011244
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
CHEBI:52991
O=[Sn]=O

Specifications

18282-10-5
SnO2
15,8905
Solubility in water: insoluble. Other solubilities: slowly soluble in sodium hydroxide solution, insoluble in alcohol and cold acids
O=[Sn]=O
150.71
CHEBI:52991
98.5% min. (metals basis)
6.95
Tin(IV) oxide
O2Sn
MFCD00011244
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
XOLBLPGZBRYERU-UHFFFAOYSA-N
dioxotin
29011
150.69
Pure
6.9500g/mL
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 242-159-

Product Identifier
  • Tin(IV) oxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.