missing translation for 'onlineSavingsMsg'
Learn More

Tin oxide, antimony doped, polymeric precursor, Oxide ≈ 30 wt%, Thermo Scientific Chemicals

Used in transparent electrical conducting electrodes

£387.93 - £786.00

Chemical Identifiers

CAS 1332-29-2
MDL Number MFCD00011244
Synonym Antimony doped tin oxide
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11354727
View Documents
Thermo Scientific Alfa Aesar
039754.14
25 g
£387.93
25g
Please to purchase this item. Need a web account? Register with us today!
11364727
View Documents
Thermo Scientific Alfa Aesar
039754.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Used in transparent electrical conducting electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1332-29-2
MFCD00011244
Antimony doped tin oxide

Specifications

1332-29-2
MFCD00011244
Soluble in alcohols
1.61 g/mL
Viscous Liquid
Antimony doped tin oxide
Oxide ≈30 wt%
Tin oxide, Antimony doped, Polymeric precursor
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'tsca' : Yes

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only