missing translation for 'onlineSavingsMsg'
Learn More

Tin oxide, polymeric precursor, Oxide ≈29 wt%, Thermo Scientific™

Product Code. 11334727
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in chemical sensors and transparent electrical conducting electrodes

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name dioxotin
SMILES O=[Sn]=O

Specifications

Physical Form Viscous Liquid
Quantity 100 g
UN Number UN1993
Solubility Information Soluble in alcohols
Concentration or Composition (by Analyte or Components) Oxide ≈29 wt%
Density 1.59 g/mL
Chemical Name or Material Tin oxide, Polymeric precursor
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'