missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin rod, 6.35cm (2.5 in.) dia., 99.85% (metals basis), Thermo Scientific™
£473.00
Chemical Identifiers
CAS | 7440-31-5 |
---|---|
Molecular Formula | Sn |
Molecular Weight (g/mol) | 118.71 |
MDL Number | MFCD00133862 |
InChI Key | ATJFFYVFTNAWJD-UHFFFAOYSA-N |
Synonym | powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german |
PubChem CID | 5352426 |
ChEBI | CHEBI:32990 |
IUPAC Name | tin |
SMILES | [Sn] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7440-31-5 | |
118.71 | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
5352426 | |
tin |
Sn | |
MFCD00133862 | |
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn germanShow More | |
CHEBI:32990 | |
[Sn] |
Specifications
7440-31-5 | |
MFCD00133862 | |
ATJFFYVFTNAWJD-UHFFFAOYSA-N | |
tin | |
5352426 | |
99.85% | |
≈2.2cm/500g | |
(metals basis) |
Sn | |
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german | |
[Sn] | |
118.71 | |
CHEBI:32990 | |
Odorless | |
Rod | |
Tin rod, 6.35cm (2.5 in.) dia. |
Safety and Handling
missing translation for 'einecsNumber' : 231-141-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tin rod, 6.35cm (2.5 in.) dia., 99.85% (metals basis), Thermo Scientific™