missing translation for 'onlineSavingsMsg'
Learn More
Tin shot, 3.175mm (0.125in) & down, 99% (metals basis), Thermo Scientific Chemicals Product Code.: 15446745

Tin shot, 3.175mm (0.125in) & down, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 15446745
1 kg
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 15446745

Brand: Thermo Scientific Alfa Aesar 045096.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Silver to Gray
Appearance Silver Gray shot
Assay Percent Range 99% (Metals basis)
Quantity 1 kg
Odor Odorless
Form Shot
Chemical Name or Material Tin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL20X011 Certificate TypeCertificate of Analysis Date27/12/2023 Catalog Number045096.22,045096.A1,045096.36
Lot NumberM19J008 Certificate TypeCertificate of Analysis Date26/01/2023 Catalog Number045096.22,045096.A1,045096.36,045098.18
Lot NumberU01F020 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number045096.22,045096.A1,045096.36,045098.18
Lot NumberJ29T009 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number045096.22,045096.A1,045096.36,045098.18
Lot NumberU10D044 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number045096.22,045096.A1,045096.36,045098.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin shot, 3.175mm (0.125in) & down, 99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.