Learn More
Lovibond™ Tintometer™ Ammonia Conditioning Reagent, 0.02 to 1mg/L
For use with Lovibond test kits, colorimeters and spectrophotometers
Brand: Lovibond™ 460170
Code : NEW
Additional Details : Weight : 0.30000kg
Description
For use with Lovibond test kits, colorimeters and spectrophotometers
- Ammonia conditioning powder for sea water
- 15g
- Range: 0.02 to 1mg/L
- Parameter: Ammonia
- Method: Indophenol blue
- For use in test kits, colorimeters and spectrophotometers
- Case of 100
Specifications
Ammonia Test | |
100 | |
0.02 to 1mg/L | |
Ammonia conditioning powder for sea water |
Test kits, colorimeters and spectrophotometers | |
15 g | |
Indophenol blue |
Safety and Handling
- 100TEST AMMONIA CONDITIONING POWDER
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : potassium tartrate dibasic hemihydrate,lithium hydroxide monohydrate