Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator, Ammonia (Powder Pack) Test, 0 to 0.5mg/L
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
Brand: Lovibond™ 146211
Code : NEW
Additional Details : Weight : 0.02900kg
Description
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
- The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
- Suitable for both mobile and static water analysis
- Available with a number of different color scales
- Provides the basis for a comprehensive easy to to to use analysis system
- Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
- Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
- The Ammonia disc supplied measures from 0 to 0.5mg/L with the following steps:
- 0, 0.05, 0.1, 0.15, 0.2, 0.25, 0.3, 0.35, 0.4, 0.45, 0.5
Specifications
Ammonia Test (Powder Pack) | |
Checkit™ Comparator | |
0 to 0.5mg/L |
Water Quality Testing | |
Carry case with Comparator, Vials, Discs, reagents and method instructions for all tests | |
Ammonia Test Disc for Checkit™ Comparator |