Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator D55 with Mirror Optics
Product Code. 10263323
Click to view available options
:
Comprator D55 with Standard Optics
Product Code. 10263323
Brand: Lovibond™ 145010
Only null left
Add to basket
Add to basket
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
  • The Lovibond™ Checkit™ comparator D55 with Mirror Optics is a compact, lightweight colorimetric unit
  • Suitable for both mobile and static water analysis
  • Available with a number of different color scales
  • Provides the basis for a comprehensive easy to to to use analysis system
  • Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
  • Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
Specifications
Description Dia: 55mm
For Use With (Application) Water Quality Testing
For Use With (Equipment) Checkit Comprator Tests
Includes Carry case with comparator, vials, discs, reagents and method instructions for all tests
Type Checkit Comprator with Mirror Optics