missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Total Nitrogen Tube Test, 0.5 to 14mg/L and 14 to 140mg/L

Use these tests with test kits, colorimeters and spectrophotometers

Brand:  Lovibond™ 2420703

Code : NEW

Additional Details : Weight : 0.30000kg Transport : UN number : 3316 Chem class : 9 Pack group : II


Product Code. 15312055

  • £119.00 / Each
Estimated Shipment: 30-08-2024
to see stock.

Description

Description

  • 24 Nitrate Tube Tests (powder, liquid reagent)
  • Parameter: Total Nitrogen
  • Method: 2,6-Dimethylphenole
  • For use in test kits, colorimeters and spectrophotometers
  • Includes:
  • Calibration Tube - 2 ct.
  • Nitrate-111 Solution - 7mL
  • Digestion Reagent Total Nitrogen - 4.5g
  • Compensation Reagent Total Nitrogen - 1.5g
Specifications

Specifications

Test Kits, Colorimeters and Spectrophotometers
24
2,6-Dimethylphenole
Calibration Tube - 2 ct., Nitrate-111 Solution - 7mL, Digestion Reagent Total Nitrogen - 4.5g, Compensation Reagent Total Nitrogen - 1.5g
0.5 to 14mg/L and 14 to 140mg/L
Total Nitrogen Tube Test
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • nitrate/total nitrogen, nitrate DMP LR, total nitrogen DMP LR/HR

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, phosphoric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers