missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ VARIO™ Molybdate HR Powder Pack Reagent Set, 25mL
Formulated for use in Lovibond test kits, colorimeters and spectrophotometers.
Brand: Lovibond™ 535400
This item is not returnable.
View return policy
Description
- Range: 0 to 35mg/L
- For use in our test kits, colorimeters and spectrophotometers
- Pre-measured, unit dose reagents
- Easy to handle, easy to open and easy to pour
- Powder pack reagents have a 5 year shelf life
- Case of 100
- Fast and easy use of VARIO Powder Packs has made them extremely popular for water testing applications in many countries throughout the world
- Provides more experienced users with a real alternative to existing measurement systems
- Produced to the same high quality standards that have made Tintometer’s tablet reagents so successful for several decades
- Parameters from aluminum and chlorine through to sulphate are just some of the well-known tests that are included in the VARIO Powder Pack range
Specifications
Water Testing | |
100 | |
0 to 35mg/L |
Test kits, colorimeters and spectrophotometers | |
25 mL | |
VARIO™ Molybdate HR Powder Pack Reagent Set, 25mL |