Learn More
Axygen™ Automation Tips for Agilent™/Velocity11 and Bravo, 10μL
GREENER_CHOICE
Compatible with automated liquid handling systems. Axygen™ Automation Tips for Agilent™/Velocity11 and Bravo, 10μL are non-filtered, nonsterile pipette tips in 384-well racks. Constructed of clear polypropyle under strict controls to ensure quality. Case includes 50 racks (19,200 tips).
Brand:  Axygen™ VT-384-10UL-SR-R
2641.78 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 13427499
  • £4221.44 / Pack of 19200
Estimated Shipment: 28-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured to stringent specifications under strict process controls
  • Assembled via automation to ensure consistent performance and quality
  • Certified RNase-/DNase-free and nonpyrogenic
  • 384-well racks of non-filtered, non-sterile pipette 10μL tips
  • Compatible with Agilent™/Velocity VPrep™ and Bravo 384ST, 96ST and 16ST head
Order Info
Minimum Order Quantity is 10 cases.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 μL
Non-sterile
DNase, RNase Free
Rack
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST
Non-filtered
Clear
Non-pyrogenic
Automation, Round
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ Automation Tips for Agilent™/Velocity11 and Bravo, 10μL > Capacity: 10μL