Learn More
BD Medical™ Tip Cap Trays
Click to view available options
For Use With (Equipment):
Conventional BD Luer Syringes
Unit Size:
Pack of 2000
Description
Enhanced productivity, reduced waste.
- Trays of ten tips packed for easy application
- Designed to minimize touch-contamination risk
- Tray packaging reduced waste compared to single-sterile caps
- 200 Trays per case (2000 tip caps)
Specifications
Specifications
| Material | Rubber |
| Product Type | Tip Tray |
| Quantity | 2000 Tips |
| Packaging | Plastic trays of 10 tips |
| For Use With (Equipment) | Conventional BD Luer Syringes |
| Connection Type | Luer |