missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP47 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10694-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
TIP47 has been described as cargo protein involved in the trafficking of the mannose-6-phosphate receptor between endosomes and the Golgi complex. Mannose 6-phophate receptors (MPRs) deliver lysosomal hydrolase from the Golgi to endosomes and then return to the Golgi complex. The protein encoded by this gene interacts with the cytoplasmic domains of both cation-independent and cation-dependent MPRs and is required for endosome to Golgi transport. This protein also binds directly to the GTPase RAB9, a member of the RAS oncogene family. The interaction with RAB9 has been shown to increase the affinity of this protein for its cargo. TIP47 has been localized in milk fat globule membranes of human and bovine origin. It has been described as a placental protein, as well. Increased amounts of TIP47 are secreted into circulation of cervix carcinoma patients. Therefore, this protein is probably significant as an oncodevelopmental marker.Specifications
TIP47 | |
Polyclonal | |
Unconjugated | |
PLIN3 | |
M6PRBP1, Perilipin 3, Placental protein 17, PLIN3, TIP47 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10226, 66905 | |
-20°C | |
Liquid |
Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60664, Q9DBG5 | |
Plin3 | |
TIP47 Fusion Protein Ag1028 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |