missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ TipTop™ Dispenser Caps for Blood Collection Tubes

Dispenses drops of filtered blood for clinical lab uses.

£230.74 - £236.94

Specifications

Includes Blood Dispenser Caps
Autoclavable Not Autoclavable
Material Polyethylene
Color Clear
No. per Pack 1000/Pk.
View More Specs

Products 2
Product Code Brand Closure Size Price Quantity & Availability  
Product Code Brand Closure Size Price Quantity & Availability  
17567956
View Documents
Labcon™
5511-800-000-9
13 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17577956
View Documents
Labcon™
5512-800-000-9
16 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Dispenser cap with integral filter for 13 or 16 mm blood collection tubes
  • Fits standard glass or plastic blood tubes
  • Polyethylene USP Class VI Resin
  • Made in USA
Specifications

Specifications

Blood Dispenser Caps
Polyethylene
1000/Pk.
ATP, RNase/Dnase, Endotoxin-free
Not Autoclavable
Clear
5000/Cs.
Non-sterile
Videos
SDS
Documents

Documents

Product Certifications