Learn More
Kartell™ Tissue Embedding Cassettes - Mega Type
Mega type for medium to large-sized samples
Brand: Kartell™ 0292503
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Specifications
White | |
40 mm | |
28 mm | |
Polyoxymethylene (POM) |