Learn More
Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, Blue
Product Code. p-4524774 Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Pack of 500
Product Code. 12648346
Brand: Epredia™ C1000AQ
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Handle small biopsies and fragile specimens with Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, the ultimate cassettes for specimen security.
  • Saves time, offers the greatest specimen security, and provides ease of use
  • Provides an inner chamber lined with a “see-through" mesh for easy identification of specimens
  • Eliminates the need for messy sponges or lens paper
  • Ensures tissues are safely processed as cassettes will not pop open during processing
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Mesh Pore Insert
Color Aqua
Product Type HistoScreen™ Cassette