Learn More
Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes
For biopsy tissue samples, these cassettes have a back-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Brand:  Epredia™ B851925YW
Additional Details : Weight : 4.53600kg
 View more versions of this product
Product Code. 16394166
  • £253.00 / Pack of 1000
Estimated Shipment: 18-09-2024
to see stock.
Description
Description
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
Small Squares
1000/Cs.
No
TissueLoc Cassette IV with Color-Matched Lids
Yes
Separate Lid, Back Hinge
Yellow